Fmoc-Tic-OH

Lieferant: Apollo Scientific

Synonyme: Fmoc-L-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, (S)-2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

OR14589-100G OR14589-25G
APOSOR14589-100GEA 197 EUR
APOSOR14589-100G APOSOR14589-25G
Fmoc-Tic-OH
Fmoc-Tic-OH
Formel: C₂₅H₂₁NO₄
Molecular Weight: 399.446 g/mol
Schmelzpunkt: 129…132 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00144368
CAS-Nummer: 136030-33-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR