Boc-3-(1-naphthyl)-L-alanine 98%

Lieferant: Apollo Scientific

Synonyme: Boc-1-Nal-OH, N-Boc-3-(1-naphthyl)-L-alanine, (S)-2-((tert-Butoxycarbonyl)amino)-3-(naphthalen-1-yl)propanoic acid

OR14594-25G OR14594-5G
APOSOR14594-25GEA 449 EUR
APOSOR14594-25G APOSOR14594-5G
Boc-3-(1-naphthyl)-L-alanine 98%
Boc-3-(1-naphthyl)-L-alanine
Formel: C₁₈H₂₁NO₄
Molecular Weight: 315.369 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00085377
CAS-Nummer: 55447-00-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR