Boc-3-(2-naphthyl)-L-alanin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Boc-3-(2-naphthyl)-L-alanine, (S)-alpha-(Boc-amino)-2-naphthalenepropionic Acid, (S)-alpha-tert-Butoxycarbonylamino-2-naphthalenepropionic Acid, Boc-2-Nal-OH
OR14598-25G
APOSOR14598-25GEA
112
EUR
APOSOR14598-25G
Boc-3-(2-naphthyl)-L-alanin
Boc-3-(2-naphthyl)-L-alanin
Formel:
C₁₈H₂₁NO₄ Molecular Weight: 315,37 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079671 CAS-Nummer: 58438-04-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...