2-Brom-6-formylpyridin 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromopyridine-2-carboxaldehyde, 6-Bromopyridine-2-carbaldehyde, 6-Bromo-2-pyridinecarboxaldehyde

OR1460-25G OR1460-100G
APOSOR1460-25GEA 55.1 EUR
APOSOR1460-25G APOSOR1460-100G
2-Brom-6-formylpyridin 95%
2-Brom-6-formylpyridin
Formel: C₆H₄BrNO
Molecular Weight: 186,01 g/mol
Schmelzpunkt: 79…85 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02683546
CAS-Nummer: 34160-40-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR