Fmoc-3-(2-naphthyl)-D-alanine 99%

Lieferant: Apollo Scientific

Synonyme: Fmoc-D-2-Nal-OH

OR14601-25G
APOSOR14601-25GEA 200 EUR
APOSOR14601-25G
Fmoc-3-(2-naphthyl)-D-alanine 99%
Fmoc-3-(2-naphthyl)-D-alanine
Formel: C₂₈H₂₃NO₄
Molecular Weight: 437,495 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077044
CAS-Nummer: 138774-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR