Boc-D-4-iodophenylalanine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Boc-D-Phe(4-I)-OH, N-Boc-4-iodo-D-phenylalanine, Boc-p-iodo-D-Phe-OH
OR14611-1G
OR14611-25G
OR14611-5G
APOSOR14611-1GEA
36.4
EUR
APOSOR14611-1G
APOSOR14611-25G
APOSOR14611-5G
Boc-D-4-iodophenylalanine
Boc-D-4-iodophenylalanine
Formel:
C₁₄H₁₈INO₄ Schmelzpunkt: 118…124 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00079670 CAS-Nummer: 176199-35-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...