N-Boc-3-(4-pyridyl)-L-alanine 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: Boc-Ala(4-pyridyl)-OH, Boc-4-Pal-OH

OR14662-1G OR14662-25G OR14662-5G
APOSOR14662-1GEA 49.3 EUR
APOSOR14662-1G APOSOR14662-25G APOSOR14662-5G
N-Boc-3-(4-pyridyl)-L-alanine 96%
N-Boc-3-(4-pyridyl)-L-alanine
Formel: C₁₃H₁₈N₂O₄
Molecular Weight: 266,297 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00672528
CAS-Nummer: 37535-57-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR