Boc-L-2-propargylglycin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-2-((tert-Butoxycarbonyl)amino)pent-4-ynoic acid, N-Boc-L-propargylglycine, (S)-N-BOC-Propargylglycine, Boc-propargyl-Gly-OH, (S)-2-(Boc-amino)-4-pentynoic Acid, Boc-Pra-OH, (S)-2-(tert-Butoxycarbonylamino)-4-pentynoic Acid

OR14674-10G OR14674-25G OR14674-5G
APOSOR14674-10GEA 288 EUR
APOSOR14674-10G APOSOR14674-25G APOSOR14674-5G
Boc-L-2-propargylglycin 97%
Boc-L-2-propargylglycin
Formel: C₁₀H₁₅NO₄
Molecular Weight: 213,23 g/mol
Schmelzpunkt: 85 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD01320855
CAS-Nummer: 63039-48-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR