Fmoc-L-propargylglycine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Fmoc-Pra-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-propargylglycine, N-Fmoc-L-propargylglycine, (S)-2-(Fmoc-amino)-4-pentynoic acid, Fmoc-propargyl-Gly-OH

OR14676-1G OR14676-5G
APOSOR14676-1GEA 29 EUR
APOSOR14676-1G APOSOR14676-5G
Fmoc-L-propargylglycine 98%
Fmoc-L-propargylglycine
Formel: C₂₀H₁₇NO₄
Molecular Weight: 335,359 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01075095
CAS-Nummer: 198561-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR