(R)-(+)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure 98%
Lieferant: Apollo Scientific
Synonyme:
(R)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic Acid, H-D-Tic-OH, D-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
OR14710-100G
OR14710-25G
APOSOR14710-100GEA
172
EUR
APOSOR14710-100G
APOSOR14710-25G
(R)-(+)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure 98%
(R)-(+)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure
Formel:
C₁₀H₁₁NO₂ Molecular Weight: 177,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144038 CAS-Nummer: 103733-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...