3-(3-Pyridyl)-L-alanine 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-2-Amino-3-(pyridin-3-yl)propanoic acid, H-β-(3-Pyridyl)-Ala-OH, H-3-Pal-OH
OR14723-1G
APOSOR14723-1GEA
45.1
EUR
APOSOR14723-1G
3-(3-Pyridyl)-L-alanine 95%
3-(3-Pyridyl)-L-alanine
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166,18 g/mol Schmelzpunkt: 248…252 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079676 CAS-Nummer: 64090-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...