H-4-Pal-OH·2HCl 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(4-Pyridyl)-alanine·2HCl, H-4-Pal-OH, L-4-Pyridylalanine

OR14725-1G OR14725-250MG OR14725-5G
APOSOR14725-1GEA 132 EUR
APOSOR14725-1G APOSOR14725-250MG APOSOR14725-5G
H-4-Pal-OH·2HCl 98%
H-4-Pal-OH·2HCl
Formel: C₈H₁₀N₂O₂·₂HCl
Molecular Weight: 239,2 g/mol
Schmelzpunkt: 257…259 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01860883
CAS-Nummer: 37535-49-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR