2-Allyl-D-glycine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: D-Allylglycine, H-D-Allyl-Gly-OH, H-D-Gly(Allyl)-OH

OR14737-5G
APOSOR14737-5GEA 109 EUR
APOSOR14737-5G
2-Allyl-D-glycine 98%
2-Allyl-D-glycine
Formel: C₅H₉NO₂
Molecular Weight: 115,1 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00063104
CAS-Nummer: 54594-06-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR