3-Brom-4'-methoxybenzophenon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Bromophenyl)(4-methoxyphenyl)methanone
OR14751-1G
OR14751-5G
APOSOR14751-1GEA
59.6
EUR
APOSOR14751-1G
APOSOR14751-5G
3-Brom-4'-methoxybenzophenon
3-Brom-4'-methoxybenzophenon
Formel:
C₁₄H₁₁BrO₂ Molecular Weight: 291,139 g/mol Schmelzpunkt: 78…80 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01311588 CAS-Nummer: 54118-76-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...