N-(tert-Butoxycarbonyl)-L-alanin

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Alpha-t-Boc-L-alanine, (S)-2-((tert-Butoxycarbonyl)amino)propanoic acid, BOC-L-Alanine, Boc-Ala-OH

OR14778-100G OR14778-250G
APOSOR14778-100GEA 21.9 EUR
APOSOR14778-100G APOSOR14778-250G
N-(tert-Butoxycarbonyl)-L-alanin
N-Boc-L-alanine
Formel: C₈H₁₅NO₄
Molecular Weight: 189,21 g/mol
Schmelzpunkt: 79…84 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00037225
CAS-Nummer: 15761-38-3
EINECS: 239-847-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR