2-Brom-5-chlorbenzoesäure
Lieferant: Apollo Scientific
|
Gefahr
|
OR1478-25G
OR1478-5G
APOSOR1478-25GEA
37.7
EUR
APOSOR1478-25G
APOSOR1478-5G
2-Brom-5-chlorbenzoesäure
2-Bromo-5-chlorobenzoic acid
Formel:
C₇H₄BrClO₂ Molecular Weight: 235,46 g/mol Schmelzpunkt: 153…157 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013982 CAS-Nummer: 21739-93-5 EINECS: 244-559-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...