tert-Butyl-2,2-dioxo-3-(3-pyridinyl)-2λ~6~-diazathiane-1-carboxylate

Lieferant: Apollo Scientific

Synonyme: 2,2-Dioxo-3-pyridin-3-yldiazathiane, N1-BOC protected

OR14827-10G OR14827-1G OR14827-5G
APOSOR14827-10GEA 894 EUR
APOSOR14827-10G APOSOR14827-1G APOSOR14827-5G
tert-Butyl-2,2-dioxo-3-(3-pyridinyl)-2λ~6~-diazathiane-1-carboxylate
tert-Butyl-2,2-dioxo-3-(3-pyridinyl)-2lambda~6~-diazathiane-1-carboxylate
Formel: C₁₀H₁₅N₃O₄S
Molecular Weight: 273,31 g/mol
Schmelzpunkt: 157…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09865023
CAS-Nummer: 1017782-68-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR