tert-Butyl-2,2-dioxo-3-(1,3-thiazol-2-yl)-2λ~6~-diazathiane-1-carboxylate

Lieferant: Apollo Scientific

Synonyme: 2,2-Dioxo-3-(1,3-thiazol-2-yl)diazathiane, N1-BOC protected

OR14828-1G OR14828-5G
APOSOR14828-1GEA 180 EUR
APOSOR14828-1G APOSOR14828-5G
tert-Butyl-2,2-dioxo-3-(1,3-thiazol-2-yl)-2λ~6~-diazathiane-1-carboxylate
tert-Butyl-2,2-dioxo-3-(1,3-thiazol-2-yl)-2lambda~6~-diazathiane-1-carboxylate
Formel: C₈H₁₃N₃O₄S₂
Molecular Weight: 279,34 g/mol
Schmelzpunkt: 143…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09865022
CAS-Nummer: 1017782-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR