1-Methylindoline-5-carboxylic acid 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR14874-1G
OR14874-5G
APOSOR14874-1GEA
0
EUR
APOSOR14874-1G
APOSOR14874-5G
1-Methylindoline-5-carboxylic acid 95+%
1-Methylindoline-5-carboxylic acid
Formel:
C₁₀H₁₁NO₂ Schmelzpunkt: 220,5…230 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09879911 CAS-Nummer: 380922-37-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...