D(-)-allo-Isoleucin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2R,3S)-(-)-α-Amino-β-methylvaleriansäure, (2R,3S)-(-)-allo-Isoleucin, (2R,3S)-(-)-2-Amino-3-methylpentansäure, H-D-allo-Ile-OH
OR14897-5G
OR14897-1G
APOSOR14897-5GEA
52.2
EUR
APOSOR14897-5G
APOSOR14897-1G
D(-)-allo-Isoleucin 97%
D(-)-allo-Isoleucin
Formel:
C₆H₁₃NO₂ Molecular Weight: 131,17 g/mol Schmelzpunkt: 285 °C Dichte: 1,235 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066445 CAS-Nummer: 1509-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...