5-Acetyl-2-brompyridin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Acetyl-6-bromopyridine

OR14930-1G OR14930-5G
APOSOR14930-1GEA 21.9 EUR
APOSOR14930-1G APOSOR14930-5G
5-Acetyl-2-brompyridin 97%
5-Acetyl-2-brompyridin
Formel: C₇H₆BrNO
Molecular Weight: 200,03 g/mol
Schmelzpunkt: 124…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04974527
CAS-Nummer: 139042-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR