Methyl-2-chlorobenzoylacetate 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-(2-chlorophenyl)-3-oxopropanoate, Methyl 3-(2-Chlorophenyl)-3-oxopropionate

OR1496-1G OR1496-5G OR1496-10G
APOSOR1496-1GEA 87 EUR
APOSOR1496-1G APOSOR1496-5G APOSOR1496-10G
Methyl-2-chlorobenzoylacetate 95+%
Methyl-2-chlorobenzoylacetate
Formel: C₁₀H₉ClO₃
Molecular Weight: 212,63 g/mol
Siedepunkt: 100…108 °C/0,15mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00216528
CAS-Nummer: 205985-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR