1H-Pyrazol-3-carbonsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Pyrazolecarboxylic acid, Pyrazole-3-carboxylic acid

OR14995-1G OR14995-5G OR14995-25G
APOSOR14995-1GEA 21.9 EUR
APOSOR14995-1G APOSOR14995-5G APOSOR14995-25G
1H-Pyrazol-3-carbonsäure
1H-Pyrazole-3-carboxylic acid
Formel: C₄H₄N₂O₂
Molecular Weight: 112.09 g/mol
Schmelzpunkt: 210…216 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00077436
CAS-Nummer: 1621-91-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR