ω-Brom-2,5-dihydroxyacetophenon

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Brom-2',5'-dihydroxyacetophenon, 2-Brom-1-(2,5-dihydroxyphenyl)ethan-1-on, α-Brom-2,5-dihydroxyacetophenon, 2,5-Dihydroxyphenacylbromid

OR15000-1G OR15000-250MG OR15000-5G
APOSOR15000-1GEA 174 EUR
APOSOR15000-1G APOSOR15000-250MG APOSOR15000-5G
ω-Brom-2,5-dihydroxyacetophenon
ω-Brom-2,5-dihydroxyacetophenon
Formel: C₈H₇BrO₃
Molecular Weight: 231,05 g/mol
Schmelzpunkt: 112…115 °C
Lagertemperatur: Kühlschrank
CAS-Nummer: 25015-91-2
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR