2-Bromo-6-methylpyrazine 97%

Lieferant: Apollo Scientific
Achtung

OR15020-1G OR15020-250MG
APOSOR15020-1GEA 52.2 EUR
APOSOR15020-1G APOSOR15020-250MG
2-Bromo-6-methylpyrazine 97%
2-Bromo-6-methylpyrazine
Formel: C₅H₅BrN₂
Molecular Weight: 173.02 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08705760
CAS-Nummer: 914452-71-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR