4-Chlor-2-nitrobenzonitril

Lieferant: Apollo Scientific
Achtung

OR15042-100G OR15042-250G OR15042-25G
APOSOR15042-100GEA 42.2 EUR
APOSOR15042-100G APOSOR15042-250G APOSOR15042-25G
4-Chlor-2-nitrobenzonitril
4-Chlor-2-nitrobenzonitril
Formel: C₇H₃ClN₂O₂
Molecular Weight: 182,57 g/mol
Schmelzpunkt: 99…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00027398
CAS-Nummer: 34662-32-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR