2-Methyl-5-nitro-2H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
OR15048-1G
OR15048-5G
APOSOR15048-1GEA
23.2
EUR
APOSOR15048-1G
APOSOR15048-5G
2-Methyl-5-nitro-2H-indazol
2-Methyl-5-nitro-2H-indazole
Formel:
C₈H₇N₃O₂ Molecular Weight: 177,16 g/mol Schmelzpunkt: 126…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051809 CAS-Nummer: 5228-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...