2-(Acetylamino)-3-[(4-bromophenyl)sulphanyl]propanoic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Acetamido)-3-[(4-bromophenyl)thio]propanoic acid

OR15059-1G OR15059-250MG
APOSOR15059-1GEA 279 EUR
APOSOR15059-1G APOSOR15059-250MG
2-(Acetylamino)-3-[(4-bromophenyl)sulphanyl]propanoic acid 95%
2-(Acetylamino)-3-[(4-bromophenyl)sulphanyl]propanoic acid
Formel: C₁₁H₁₂BrNO₃S
Molecular Weight: 318,2 g/mol
Schmelzpunkt: 152…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00869819
CAS-Nummer: 126253-78-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR