3-(1,3-Benzoxazol-2-yl)-2-pyridinol
Lieferant: Apollo Scientific
Synonyme:
3-(1,3-Benzoxazol-2-yl)-2-hydroxypyridine
OR15065-10G
OR15065-1G
OR15065-5G
APOSOR15065-10GEA
894
EUR
APOSOR15065-10G
APOSOR15065-1G
APOSOR15065-5G
3-(1,3-Benzoxazol-2-yl)-2-pyridinol
3-(1,3-Benzoxazol-2-yl)-2-pyridinol
Formel:
C₁₂H₈N₂O₂ Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08444020 CAS-Nummer: 292140-74-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...