3-Bromo-1H-indole-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR15068-1G OR15068-5G
APOSOR15068-1GEA 39.3 EUR
APOSOR15068-1G APOSOR15068-5G
3-Bromo-1H-indole-2-carbohydrazide
3-Bromo-1H-indole-2-carbohydrazide
Formel: C₉H₈BrN₃O
Molecular Weight: 254.09 g/mol
Schmelzpunkt: 192…194 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08444014
CAS-Nummer: 441801-35-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR