2,3-Dihydro-1-benzofuran-5-sulphonamide
Lieferant: Apollo Scientific
|
Achtung
|
OR15079-1G
OR15079-5G
OR15079-250MG
APOSOR15079-1GEA
132
EUR
APOSOR15079-1G
APOSOR15079-5G
APOSOR15079-250MG
2,3-Dihydro-1-benzofuran-5-sulphonamide
2,3-Dihydro-1-benzofuran-5-sulphonamide
Formel:
C₈H₉NO₃S Molecular Weight: 199,23 g/mol Schmelzpunkt: 164…166 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03069322 CAS-Nummer: 112894-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...