4-Phthalimidobutyric acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(1,3-Dioxoisoindolin-2-yl)butanoic acid, 4-(Phthalimid-1-yl)butanoic acid

OR15083-1G OR15083-250MG
APOSOR15083-1GEA 63.8 EUR
APOSOR15083-1G APOSOR15083-250MG
4-Phthalimidobutyric acid
4-Phthalimidobutyric acid
Formel: C₁₂H₁₁NO₄
Molecular Weight: 233.22 g/mol
Schmelzpunkt: 119…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00196079
CAS-Nummer: 3130-75-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR