Ethyl-6-brom-4-hydroxychinolin-3-carboxylat 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 6-bromo-4-hydroxyquinoline-3-carboxylate

OR15084-25G
APOSOR15084-25GEA 171 EUR
APOSOR15084-25G
Ethyl-6-brom-4-hydroxychinolin-3-carboxylat 95%
Ethyl-6-brom-4-hydroxychinolin-3-carboxylat
Formel: C₁₂H₁₀BrNO₃
Molecular Weight: 296,1200 g/mol
Schmelzpunkt: 326…329 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173362
CAS-Nummer: 122794-99-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR