Ethyl-2-bromo-3-oxo-3-phenylpropanoate

Lieferant: Apollo Scientific
Achtung

OR15085-1G OR15085-5G
APOSOR15085-1GEA 180 EUR
APOSOR15085-1G APOSOR15085-5G
Ethyl-2-bromo-3-oxo-3-phenylpropanoate
Ethyl-2-bromo-3-oxo-3-phenylpropanoate
Formel: C₁₁H₁₁BrO₃
Molecular Weight: 271,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08444026
CAS-Nummer: 55919-47-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR