2-Methyl-1,3-benzothiazole-6-carboxylic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR15100-1G
OR15100-250MG
OR15100-5G
APOSOR15100-1GEA
174
EUR
APOSOR15100-1G
APOSOR15100-250MG
APOSOR15100-5G
2-Methyl-1,3-benzothiazole-6-carboxylic acid 95%
2-Methyl-1,3-benzothiazole-6-carboxylic acid
Formel:
C₉H₇NO₂S Schmelzpunkt: 229…238 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07394023 CAS-Nummer: 6941-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...