Methyl-3-bromo-1-(cyanomethyl)-1H-indole-2-carboxylate

Lieferant: Apollo Scientific

OR15101-10G OR15101-1G OR15101-5G
APOSOR15101-10GEA 1690 EUR
APOSOR15101-10G APOSOR15101-1G APOSOR15101-5G
Methyl-3-bromo-1-(cyanomethyl)-1H-indole-2-carboxylate
Methyl-3-bromo-1-(cyanomethyl)-1H-indole-2-carboxylate
Formel: C₁₂H₉BrN₂O₂
Molecular Weight: 293.13 g/mol
Schmelzpunkt: 170…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08444032
CAS-Nummer: 924869-15-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR