Methyl-3-iodo-1H-indole-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR15108-250MG
OR15108-500MG
APOSOR15108-250MGEA
39.3
EUR
APOSOR15108-250MG
APOSOR15108-500MG
Methyl-3-iodo-1H-indole-2-carboxylate
Methyl-3-iodo-1H-indole-2-carboxylate
Formel:
C₁₀H₈INO₂ Molecular Weight: 301,09 g/mol Schmelzpunkt: 157…159 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04124152 CAS-Nummer: 534595-85-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...