2-Aminobenzo[d]thiazole-6-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Aminobenzothiazole-6-carboxylic acid

OR15130-5G
APOSOR15130-5GEA 58 EUR
APOSOR15130-5G
2-Aminobenzo[d]thiazole-6-carboxylic acid
2-Aminobenzo[d]thiazole-6-carboxylic acid
Formel: C₈H₆N₂O₂S
Molecular Weight: 194,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00054180
CAS-Nummer: 93-85-6
EINECS: 202-283-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR