N-(3-Bromophenyl)-N'-(2-chloroethyl)urea

Lieferant: Apollo Scientific

OR15136-10G OR15136-1G OR15136-5G
APOSOR15136-10GEA 0 EUR
APOSOR15136-10G APOSOR15136-1G APOSOR15136-5G
N-(3-Bromophenyl)-N'-(2-chloroethyl)urea
N-(3-Bromophenyl)-N'-(2-chloroethyl)urea
Formel: C₉H₁₀BrClN₂O
Schmelzpunkt: 108…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00451942
CAS-Nummer: 146257-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR