1-(3-Bromophenyl)tetrahydro-2H-imidazol-2-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(3-Bromophenyl)imidazolidin-2-one
OR15138-100MG
OR15138-1G
OR15138-250MG
APOSOR15138-100MGEA
21.9
EUR
APOSOR15138-100MG
APOSOR15138-1G
APOSOR15138-250MG
1-(3-Bromophenyl)tetrahydro-2H-imidazol-2-one
1-(3-Bromophenyl)tetrahydro-2H-imidazol-2-one
Formel:
C₉H₉BrN₂O Molecular Weight: 241,09 g/mol Schmelzpunkt: 132…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08443974 CAS-Nummer: 14088-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...