N-(4-Bromophenyl)-N'-(2-chloroethyl)urea

Lieferant: Apollo Scientific

OR15141-10G OR15141-1G OR15141-5G
APOSOR15141-10GEA 894 EUR
APOSOR15141-10G APOSOR15141-1G APOSOR15141-5G
N-(4-Bromophenyl)-N'-(2-chloroethyl)urea
N-(4-Bromophenyl)-N'-(2-chloroethyl)urea
Formel: C₉H₁₀BrClN₂O
Schmelzpunkt: 172…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08444007
CAS-Nummer: 15145-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR