1-(4-Bromophenyl)tetrahydro-2H-imidazol-2-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Bromophenyl)imidazolidin-2-one

OR15143-1G OR15143-5G
APOSOR15143-1GEA 105 EUR
APOSOR15143-1G APOSOR15143-5G
1-(4-Bromophenyl)tetrahydro-2H-imidazol-2-one
1-(4-Bromophenyl)tetrahydro-2H-imidazol-2-one
Formel: C₉H₉BrN₂O
Molecular Weight: 241,09 g/mol
Schmelzpunkt: 189…191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08444010
CAS-Nummer: 530081-14-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR