2-Chlor-6-nitrobenzothiazol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-6-nitrobenzo[d]thiazole, 2-Chloro-6-nitro-1,3-benzothiazole
OR15153-1G
OR15153-5G
APOSOR15153-1GEA
21.9
EUR
APOSOR15153-1G
APOSOR15153-5G
2-Chlor-6-nitrobenzothiazol
2-Chloro-6-nitrobenzothiazole
Formel:
C₇H₃ClN₂O₂S Schmelzpunkt: 189…195 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00022852 CAS-Nummer: 2407-11-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...