3-(3-Chlorphenyl)-1H-pyrazol-5-amin

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-3-(3-chlorophenyl)-1H-pyrazole

OR15158-1G OR15158-5G
APOSOR15158-1GEA 24.8 EUR
APOSOR15158-1G APOSOR15158-5G
3-(3-Chlorphenyl)-1H-pyrazol-5-amin
3-(3-Chlorphenyl)-1H-pyrazol-5-amin
Formel: C₉H₈ClN₃
Molecular Weight: 193,6329 g/mol
Schmelzpunkt: 138…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02664265
CAS-Nummer: 208519-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR