2-(4-Chlorophenoxy)nicotinaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR15159-1G
OR15159-5G
APOSOR15159-1GEA
244
EUR
APOSOR15159-1G
APOSOR15159-5G
2-(4-Chlorophenoxy)nicotinaldehyde 95%
2-(4-Chlorophenoxy)nicotinaldehyde
Formel:
C₁₂H₈ClNO₂ Schmelzpunkt: 90…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00794360 CAS-Nummer: 478031-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...