3',4'-Dichloracetanilid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(3,4-Dichlorophenyl)acetamide
OR15167-25G
APOSOR15167-25GEA
255
EUR
APOSOR15167-25G
3',4'-Dichloracetanilid
3',4'-Dichloracetanilid
Formel:
C₈H₇Cl₂NO Molecular Weight: 204,0500 g/mol Schmelzpunkt: 121…124 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00018148 CAS-Nummer: 2150-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...