Ethyl-2-chloro-6-benzothiazolecarboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-chloro-6-benzothiazolecarboxylate

OR15184-5G
APOSOR15184-5GEA 98.6 EUR
APOSOR15184-5G
Ethyl-2-chloro-6-benzothiazolecarboxylate
Ethyl-2-chloro-6-benzothiazolecarboxylate
Formel: C₁₀H₈ClNO₂S
Molecular Weight: 241.6900 g/mol
Schmelzpunkt: 90…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08443995
CAS-Nummer: 78485-37-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR