5-Hydroxy-2(1H)-quinolinone 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Hydroxyquinolin-2(1H)-one
OR15208-1G
OR15208-250MG
OR15208-100MG
APOSOR15208-1GEA
48
EUR
APOSOR15208-1G
APOSOR15208-250MG
APOSOR15208-100MG
5-Hydroxy-2(1H)-quinolinone 99%
5-Hydroxy-2(1H)-quinolinone
Formel:
C₉H₇NO₂ Molecular Weight: 161,16 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02181132 CAS-Nummer: 31570-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...