Methyl-4-amino-3-brom-5-nitrobenzoat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-amino-3-bromo-5-nitrobenzoate
OR15218-1G
OR15218-25G
OR15218-5G
APOSOR15218-1GEA
21.9
EUR
APOSOR15218-1G
APOSOR15218-25G
APOSOR15218-5G
Methyl-4-amino-3-brom-5-nitrobenzoat
Methyl-4-amino-3-brom-5-nitrobenzoat
Formel:
C₈H₇BrN₂O₄ Molecular Weight: 275,06 g/mol Schmelzpunkt: 108…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08443938 CAS-Nummer: 105655-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...