Methyl-4-({[(2-chloroethyl)amino]carbonyl}amino)benzenecarboxylate

Lieferant: Apollo Scientific

OR15220-10G OR15220-1G OR15220-5G
APOSOR15220-10GEA 1350 EUR
APOSOR15220-10G APOSOR15220-1G APOSOR15220-5G
Methyl-4-({[(2-chloroethyl)amino]carbonyl}amino)benzenecarboxylate
Methyl-4-({[(2-chloroethyl)amino]carbonyl}amino)benzenecarboxylate
Formel: C₁₁H₁₃ClN₂O₃
Molecular Weight: 256,69 g/mol
Schmelzpunkt: 165…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03149047
CAS-Nummer: 380324-25-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR